3-Methyl-1H-indazol-6-amine 95%

Supplier: Ambeed
Warning

A371430-10G A371430-1G A371430-250MG A371430-25G A371430-5G
77033-296EA 319.34 USD
77033-296 77033-298 77033-300 77033-302 77033-304
3-Methyl-1H-indazol-6-amine 95%
3-Methyl-1H-indazol-6-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.18 g/mol
MDL Number: MFCD11007871
CAS Number: 79173-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR