N-(6-Ethoxy-2-benzothiazolyl)benzenesulfonamide 95%

Supplier: Ambeed
Warning

A371332-100MG A371332-1G A371332-250MG
77015-994EA 35.01 USD
77015-994 77015-996 77015-998
N-(6-Ethoxy-2-benzothiazolyl)benzenesulfonamide 95%
N-(6-Ethoxy-2-benzothiazolyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₃S₂
MW: 334.41 g/mol
MDL Number: MFCD02805711
CAS Number: 15850-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR