7-Methoxy-1-naphthylacetonitrile 97%

Supplier: Ambeed
Warning

A371135-10G A371135-1G A371135-250MG A371135-5G
77033-288EA 29 USD
77033-288 77033-290 77033-292 77033-294
7-Methoxy-1-naphthylacetonitrile 97%
7-Methoxy-1-naphthylacetonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MDL Number: MFCD08704309
CAS Number: 138113-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR