2-(4-Bromophenyl)-5-methyl-1,3,4-oxadiazole ≥98%

Supplier: Ambeed
Warning

A370971-100MG A370971-1G A370971-250MG
77015-988EA 44.56 USD
77015-988 77015-990 77015-992
2-(4-Bromophenyl)-5-methyl-1,3,4-oxadiazole ≥98%
2-(4-Bromophenyl)-5-methyl-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MW: 239.07 g/mol
MDL Number: MFCD00491620
CAS Number: 41421-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR