2-Bromo-3-(trifluoromethyl)phenol 98%
Supplier: Ambeed
|
Warning
|
A370968-100MG
A370968-1G
A370968-250MG
A370968-5G
76989-882EA
41
USD
76989-882
76989-884
76989-886
76989-888
2-Bromo-3-(trifluoromethyl)phenol 98%
2-Bromo-3-(trifluoromethyl)phenol
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃O |
MDL Number:
MFCD13194487 CAS Number: 1214323-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...