3-Ethynylimidazo[1,2-b]pyridazine 98%

Supplier: Ambeed
Warning

A370943-5G A370943-1G
77015-986EA 101 USD
77015-986 77568-760
3-Ethynylimidazo[1,2-b]pyridazine 98%
3-Ethynylimidazo[1,2-b]pyridazine

Precaución: For research use only.

Formula: C₈H₅N₃
MDL Number: MFCD17012027
CAS Number: 943320-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR