1-Difluoromethyl-1H-pyrazole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(Difluoromethyl)-1H-pyrazole-3-carboxylic acid
A370860-100MG
A370860-250MG
A370860-1G
77003-494EA
55.17
USD
77003-494
77003-496
77672-860
1-Difluoromethyl-1H-pyrazole-3-carboxylic acid 98%
1-Difluoromethyl-1H-pyrazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₄F₂N₂O₂ MW: 162.1 g/mol |
MDL Number:
MFCD03420211 CAS Number: 925179-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...