3,3-Dimethoxycyclobutanecarboxylic acid 95%

Supplier: Ambeed
Warning

A370337-1G A370337-25G
77033-276EA 16.98 USD
77033-276 77033-278
3,3-Dimethoxycyclobutanecarboxylic acid 95%
3,3-Dimethoxycyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.16 g/mol
MDL Number: MFCD12755812
CAS Number: 332187-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR