3-Quinolineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-3-boronic acid pinacolate, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, Quinoline-3-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-quinolyl)-1,3,2-dioxaborolane

A370279-1G A370279-25G A370279-5G
77003-488EA 19.1 USD
77003-488 77003-490 77003-492
3-Quinolineboronic acid pinacol ester 98%
3-Quinolineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₂
MW: 255.1199 g/mol
MDL Number: MFCD05155218
CAS Number: 171364-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR