1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)ethanone 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Acetylpyridine-5-boronic acid pinacol ester
A370214-100MG
A370214-1G
A370214-250MG
A370214-5G
77033-268EA
37.14
USD
77033-268
77033-270
77033-272
77033-274
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)ethanone 98+%
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BNO₃ MW: 247.10 g/mol |
MDL Number:
MFCD12923434 CAS Number: 741709-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...