2-(Pyridin-4-yl)-1-(p-tolyl)ethanone 98%

Supplier: Ambeed
Warning

A370138-1G A370138-250MG
77015-974EA 62.6 USD
77015-974 77015-976
2-(Pyridin-4-yl)-1-(p-tolyl)ethanone 98%
2-(Pyridin-4-yl)-1-(p-tolyl)ethanone

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO
MW: 211.26 g/mol
MDL Number: MFCD04114381
CAS Number: 100866-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR