6-(Hydroxymethyl)-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: (1H-Indazol-6-yl)methanol

A370044-1G A370044-250MG A370044-5G
77033-264EA 72.15 USD
77033-264 77033-266 77672-852
6-(Hydroxymethyl)-1H-indazole 95%
6-(Hydroxymethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD11052639
CAS Number: 916902-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR