4-Aminobenzonitrile 98%

Supplier: Ambeed
Warning

A369331-100G A369331-10G A369331-25G A369331-500G A369331-1000G
77033-256EA 44 USD
77033-256 77033-258 77033-260 77033-262 77601-630
4-Aminobenzonitrile 98%
4-Aminobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂
MDL Number: MFCD00007821
CAS Number: 873-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR