Brivanib alaninate ≥98%

Supplier: Ambeed
Danger

Synonyms: BMS-582664, (R)-(S)-1-((4-((4-Fluoro-2-methyl-1H-indol-5-yl)oxy)-5-methylpyrrolo[2,1-f][1,2,4]triazin-6-yl)oxy)propan-2-yl 2-aminopropanoate, Brivanib (alaninate)

A369272-1MG A369272-2MG A369272-5MG
76989-864EA 30.77 USD
76989-864 76989-866 76989-868
Brivanib alaninate ≥98%
Brivanib alaninate

Precaución: for Research Use Only

Formula: C₂₂H₂₄FN₅O₄
MW: 441.5 g/mol
MDL Number: MFCD22577162
CAS Number: 649735-63-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR