6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-6-carboxylate

A368555-100MG A368555-1G A368555-250MG A368555-5G
77015-938EA 43 USD
77015-938 77015-940 77015-942 77015-944
6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 96%
6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₀BNO₄
MW: 301.1453 g/mol
MDL Number: MFCD11858368
CAS Number: 1256359-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR