4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(trifluoromethyl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-(trifluoromethyl)-1H-pyrazole-4-boronic acid, pinacol ester

A368415-100MG A368415-1G A368415-250MG A368415-5G
76989-858EA 18 USD
76989-858 77885-967 77885-968 77885-969
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(trifluoromethyl)-1H-pyrazole 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₁₄BF₃N₂O₂
MDL Number: MFCD18383267
CAS Number: 1046831-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR