2,6-Dichloro-4-methylpyridin-3-amine 98%

Supplier: Ambeed
Warning

A368218-10G A368218-250MG A368218-25G A368218-5G A368218-100G
77033-244EA 13 USD
77033-244 77033-246 77033-248 77033-250 77872-568
2,6-Dichloro-4-methylpyridin-3-amine 98%
2,6-Dichloro-4-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MDL Number: MFCD01320814
CAS Number: 129432-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR