3'-Bromobiphenyl-3-carboxylic acid 95%

Supplier: Ambeed
Danger

A368144-10G A368144-250MG A368144-5G
77033-238EA 419.06 USD
77033-238 77033-240 77033-242
3'-Bromobiphenyl-3-carboxylic acid 95%
3'-Bromobiphenyl-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉BrO₂
MW: 277.12 g/mol
MDL Number: MFCD00093210
CAS Number: 854237-06-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR