7-Nitro-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole

A367353-100G A367353-100MG A367353-10G A367353-1G A367353-250MG A367353-25G A367353-5G
77003-470EA 219 USD
77003-470 77003-472 77003-474 77003-476 77003-478 77003-480 77003-482
7-Nitro-1H-indazole 98%
7-Nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD00022789
CAS Number: 2942-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR