4-Bromo-7-chlorobenzofuran 95%

Supplier: Ambeed
Warning

A367159-100MG A367159-250MG
76989-852EA 44.56 USD
76989-852 77613-084
4-Bromo-7-chlorobenzofuran 95%
4-Bromo-7-chlorobenzofuran

Precaución: for Research Use Only

Formula: C₈H₄BrClO
MW: 231.47 g/mol
MDL Number: MFCD11877827
CAS Number: 855343-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR