2-Chloro-3-methyl-5-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-methylpyridine-5-boronic Acid Pinacol Ester

A366854-1G A366854-250MG A366854-5G
77033-232EA 35 USD
77033-232 77033-234 77033-236
2-Chloro-3-methyl-5-pyridineboronic acid pinacol ester 98%
2-Chloro-3-methyl-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
MDL Number: MFCD08063084
CAS Number: 1010101-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR