4,4',4''-((1,3,5-Triazine-2,4,6-triyl)tris(azanediyl))tribenzoic acid 95%

Supplier: Ambeed
Warning

A366717-1G A366717-250MG A366717-5G
77015-932EA 17 USD
77015-932 77015-934 77015-936
4,4',4''-((1,3,5-Triazine-2,4,6-triyl)tris(azanediyl))tribenzoic acid 95%
4,4',4''-((1,3,5-Triazine-2,4,6-triyl)tris(azanediyl))tribenzoic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₈N₆O₆
MDL Number: MFCD01232264
CAS Number: 63557-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR