2-Nitro-α,α,α-trifluoro-p-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Nitro-4-(trifluoromethyl)benzoic acid, 2-Nitro-4-trifluoromethylbenzoic acid

A366673-100G A366673-10G A366673-25G A366673-500G A366673-5G
77003-460EA 83 USD
77003-460 77003-462 77003-464 77003-466 77003-468
2-Nitro-α,α,α-trifluoro-p-toluic acid 97%
2-Nitro-α,α,α-trifluoro-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₄
MDL Number: MFCD00007142
CAS Number: 320-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR