4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride 95%

Supplier: Ambeed
Warning

A366642-100MG A366642-250MG A366642-5G
77015-926EA 25 USD
77015-926 77015-928 77015-930
4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride 95%
4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₅F₆NO·ClH
MW: 281.583 g/mol
MDL Number: MFCD27923427
CAS Number: 1588441-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR