(R)-Methyl piperidine-3-carboxylate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A366113-100MG
A366113-1G
A366113-250MG
A366113-25G
77015-920EA
9
USD
77015-920
77015-922
77015-924
77890-753
(R)-Methyl piperidine-3-carboxylate hydrochloride 98%
(R)-Methyl piperidine-3-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ |
MDL Number:
MFCD08274539 CAS Number: 1255651-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...