3-Amino-4-bromo-6-chloropicolinic acid 95%

Supplier: Ambeed
Warning

A365702-100MG A365702-1G A365702-250MG A365702-50MG
77015-912EA 187.78 USD
77015-912 77015-914 77015-916 77015-918
3-Amino-4-bromo-6-chloropicolinic acid 95%
3-Amino-4-bromo-6-chloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄BrClN₂O₂
MW: 251.46 g/mol
MDL Number: MFCD18382761
CAS Number: 1073182-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR