FMOC-L-3,4-difluorophenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(3,4-DiF)-OH

A365382-1G A365382-250MG A365382-5G A365382-25G
77003-444EA 38 USD
77003-444 77003-446 77672-836 77881-994
FMOC-L-3,4-difluorophenylalanine 97%
FMOC-L-3,4-difluorophenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₁₉F₂NO₄
MDL Number: MFCD00672550
CAS Number: 198560-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR