Ethyl-2-chloroquinoline-4-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A365224-100MG
A365224-1G
A365224-250MG
77003-440EA
16
USD
77003-440
77003-442
77672-834
Ethyl-2-chloroquinoline-4-carboxylate 98%
Ethyl 2-chloroquinoline-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₈ClNO₂ |
MDL Number:
MFCD03413689 CAS Number: 62482-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...