N-Methyl-N-[(1-methyl-1H-pyrazol-4-yl)methyl]amine 95%

Supplier: Ambeed
Danger

Synonyms: N-Methyl-1-(1-methyl-1H-pyrazol-4-yl)methanamine

A365190-100MG A365190-1G A365190-250MG
77033-218EA 42 USD
77033-218 77033-220 77033-222
N-Methyl-N-[(1-methyl-1H-pyrazol-4-yl)methyl]amine 95%
N-Methyl-N-[(1-methyl-1H-pyrazol-4-yl)methyl]amine

Precaución: for Research Use Only

Formula: C₆H₁₁N₃
MW: 125.17 g/mol
MDL Number: MFCD02090919
CAS Number: 179873-43-9
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR