(S)-Ethyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate 95%

Supplier: Ambeed
Warning

A364448-10G A364448-1G
77015-906EA 50 USD
77015-906 77015-908 77015-910
(S)-Ethyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate 95%
(S)-Ethyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₅
MDL Number: MFCD15141969
CAS Number: 84787-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR