4-Bromo-2-fluoroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 4-bromo-2-fluoro-1-methoxybenzene

A364295-1000G A364295-100G A364295-10G A364295-25G A364295-500G A364295-5G
77003-420EA 304.48 USD
77003-420 77003-422 77003-424 77003-426 77003-428 77003-430
4-Bromo-2-fluoroanisole 98%
4-Bromo-2-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.03 g/mol
MDL Number: MFCD00011710
CAS Number: 2357-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR