Methyl 3,5-difluoro-4-nitrobenzoate 95%
Supplier: Ambeed
|
Danger
|
A364096-1G
A364096-5G
76989-840EA
128.37
USD
76989-840
77684-066
Methyl 3,5-difluoro-4-nitrobenzoate 95%
Methyl 3,5-difluoro-4-nitrobenzoate
Precaución: For research use only.
Formula:
C₈H₅F₂NO₄ MW: 217.12 g/mol |
MDL Number:
MFCD25955661 CAS Number: 1123172-87-1 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...