7-Deaza-7-iodo-2'-deoxyadenosine 98%

Supplier: Ambeed
Warning

Synonyms: 7-Deaza-2'-deoxy-7-iodoadenosine, (2R,3S,5R)-5-(4-Amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

A364032-100MG A364032-1G A364032-250MG A364032-5G
76989-828EA 40.32 USD
76989-828 76989-830 76989-832 77572-080
7-Deaza-7-iodo-2'-deoxyadenosine 98%
7-Deaza-7-iodo-2'-deoxyadenosine

Precaución: for Research Use Only

Formula: C₁₁H₁₃IN₄O₃
MW: 376.1504 g/mol
MDL Number: MFCD07778650
CAS Number: 166247-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR