1-(4-Bromophenylsulfonyl)pyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-N-pyrrolidin-1-ylbenzenesulphonamide, 1-((4-Bromophenyl)sulphonyl)pyrrolidine

A363986-100MG A363986-1G A363986-250MG
76989-822EA 11.67 USD
76989-822 76989-824 76989-826
1-(4-Bromophenylsulfonyl)pyrrolidine 98%
1-(4-Bromophenylsulphonyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂S
MW: 290.1768 g/mol
MDL Number: MFCD00584577
CAS Number: 136350-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR