Methyl-4-hydroxy-3-methoxy-5-nitrobenzenecarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-hydroxy-3-methoxy-5-nitrobenzoate
A363928-100MG
A363928-1G
A363928-250MG
A363928-25G
A363928-5G
77003-406EA
7
USD
77003-406
77003-408
77003-410
77003-412
77003-414
Methyl-4-hydroxy-3-methoxy-5-nitrobenzenecarboxylate 95%
Methyl-4-hydroxy-3-methoxy-5-nitrobenzenecarboxylate
Precaución: for Research Use Only
Formula:
C₉H₉NO₆ |
MDL Number:
MFCD06203667 CAS Number: 42590-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...