4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(1H-Pyrrolo[2,3-d]pyrimidin-4-yl)morpholine

A363910-100MG A363910-250MG
77015-902EA 36 USD
77015-902 77015-904
4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine 95%
4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O
MW: 204.23 g/mol
MDL Number: MFCD18904308
CAS Number: 90870-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR