3-(3,4-Dihydroxyphenyl)-2-oxopropanoic acid 97%

Supplier: Ambeed
Warning

A363781-1G A363781-250MG A363781-100MG
77003-402EA 307 USD
77003-402 77003-404 77672-830
3-(3,4-Dihydroxyphenyl)-2-oxopropanoic acid 97%
3-(3,4-Dihydroxyphenyl)-2-oxopropanoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₅
MDL Number: MFCD08460177
CAS Number: 4228-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR