Fmoc-D-3,3-diphenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-diphenylpropanoic acid

A363307-1G A363307-5G A363307-250MG
77003-398EA 59.42 USD
77003-398 77003-400 77672-828
Fmoc-D-3,3-diphenylalanine 98%
Fmoc-D-3,3-diphenylalanine

Precaución: for Research Use Only

Formula: C₃₀H₂₅NO₄
MW: 463.54 g/mol
MDL Number: MFCD00672338
CAS Number: 189937-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR