Bis(2-ethylhexyl)adipate 97%

Supplier: Ambeed
Warning

Synonyms: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate

A362872-10G A362872-25G
77033-206EA 5 USD
77033-206 77033-208
Bis(2-ethylhexyl)adipate 97%
Bis(2-ethylhexyl)adipate

Precaución: for Research Use Only

Formula: C₂₂H₄₂O₄
MDL Number: MFCD00009496
CAS Number: 103-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR