1H-Imidazol-2-ylmethylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (1H-Imidazol-2-yl)methanamine hydrochloride

A362759-100MG A362759-1G A362759-250MG
77015-890EA 47.75 USD
77015-890 77015-892 77015-894
1H-Imidazol-2-ylmethylamine hydrochloride 95%
1H-Imidazol-2-ylmethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClN₃
MW: 133.58 g/mol
MDL Number: MFCD07366514
CAS Number: 138799-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR