4-Bromo-1,5-dimethyl-1H-pyrazole 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Bromo-1,5-dimethylpyrazole
A362131-1G
A362131-250MG
A362131-5G
A362131-25G
77033-200EA
44.56
USD
77033-200
77033-202
77033-204
77568-736
4-Bromo-1,5-dimethyl-1H-pyrazole 98%
4-Bromo-1,5-dimethyl-1H-pyrazole
Precaución: For research use only.
Formula:
C₅H₇BrN₂ MW: 175.0265 g/mol |
MDL Number:
MFCD09759006 CAS Number: 5775-86-0 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...