3,5-Dichloro-4-methoxyaniline 97%

Supplier: Ambeed
Warning

A361413-1G A361413-250MG
77003-354EA 123.07 USD
77003-354 77003-356
3,5-Dichloro-4-methoxyaniline 97%
3,5-Dichloro-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.04 g/mol
MDL Number: MFCD11647707
CAS Number: 32407-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR