2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene 97%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 4,4',4''-((2,4,6-Trimethylbenzene-1,3,5-triyl)tris(methylene))tris(2,6-di-tert-butylphenol)

A361290-100G A361290-25G
77003-350EA 19 USD
77003-350 77003-352
2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene 97%
2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene

Precaución: for Research Use Only

Formula: C₅₄H₇₈O₃
MW: 775.22 g/mol
MDL Number: MFCD00026284
CAS Number: 1709-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR