2-Amino-4-hydroxypyridine hydrochloride 98%

Supplier: Ambeed
Warning

A361247-1G A361247-5G
77033-196EA 29.71 USD
77033-196 77033-198
2-Amino-4-hydroxypyridine hydrochloride 98%
2-Amino-4-hydroxypyridine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇ClN₂O
MW: 146.57 g/mol
MDL Number: MFCD09701349
CAS Number: 1187932-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR