3-Benzyloxy-6-bromo-2-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Benzyloxy)-6-bromo-2-fluorophenylboronic acid

A361014-1G A361014-5G
77033-192EA 13.8 USD
77033-192 77033-194
3-Benzyloxy-6-bromo-2-fluorophenylboronic acid 97%
3-Benzyloxy-6-bromo-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BBrFO₃
MW: 324.9400 g/mol
MDL Number: MFCD09475868
CAS Number: 957035-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR