tert-Butyl (4R-cis)-6-formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3R,5S)-6-oxo-3,5-o-iso-propylidene-3,5-dihydroxyhexanoate, tert-Butyl (4R-cis)-6-formaldehydel-2,2-dimethyl-1,3-dioxane-4-acetate, (4R-cis)-6-Formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetic acid 1,1-dimethylethyl ester

A361005-1G A361005-250MG A361005-25G A361005-10G A361005-5G
77033-188EA 6 USD
77033-188 77033-190 77551-036 77872-547 77872-548
tert-Butyl (4R-cis)-6-formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetate 95%
tert-Butyl (4R-cis)-6-formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetate

Precaución: for Research Use Only

Formula: C₁₃H₂₂O₅
MDL Number: MFCD02093270
CAS Number: 124752-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR