Ethyl-3-bromo-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromo-5-nitrobenzoate, 3-Bromobenzoic acid ethyl ester

A360393-250MG A360393-5G
77003-346EA 15.92 USD
77003-346 77003-348
Ethyl-3-bromo-5-nitrobenzoate 98%
Ethyl-3-bromo-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MW: 274.07 g/mol
MDL Number: MFCD09258760
CAS Number: 690260-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR