N-Boc-3-bromo-4-fluoroaniline 97%
Supplier: Ambeed
|
Danger
|
A360279-100MG
A360279-1G
A360279-250MG
A360279-5G
A360279-10G
77015-862EA
18.04
USD
77015-862
77015-864
77015-866
77015-868
77684-058
N-Boc-3-bromo-4-fluoroaniline 97%
N-Boc-3-bromo-4-fluoroaniline
Precaución: For research use only.
Formula:
C₁₁H₁₃BrFNO₂ MW: 290.1300 g/mol |
MDL Number:
MFCD11975628 CAS Number: 836619-77-3 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...