3-(Bromomethyl)-5-methylpyridine hydrobromide 97%

Supplier: Ambeed
Danger

A360128-1G A360128-250MG A360128-5G
77033-180EA 50.93 USD
77033-180 77033-182 77033-184
3-(Bromomethyl)-5-methylpyridine hydrobromide 97%
3-(Bromomethyl)-5-methylpyridine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₉Br₂N
MW: 266.96 g/mol
MDL Number: MFCD20921819
CAS Number: 1235342-53-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR