4-Chloro-5-hydroxypyridazin-3(2H)-one 95%

Supplier: Ambeed
Warning

A359586-1G A359586-250MG A359586-5G
77033-174EA 54 USD
77033-174 77033-176 77033-178
4-Chloro-5-hydroxypyridazin-3(2H)-one 95%
4-Chloro-5-hydroxypyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₄H₃ClN₂O₂
MDL Number: MFCD00832513
CAS Number: 64178-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR